Product Details

SNP ID
rs7660054
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:55402340 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGAAAATTAAAATACTTTAATGT[C/T]ATCTTTCGAAATCAGGTTTATCTGT
Phenotype
MIM: 614726
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM165 PubMed Links

Gene Details

Gene
TMEM165
Gene Name
transmembrane protein 165
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018475.4 Intron NP_060945.2
XM_011534394.2 Intron XP_011532696.1
XM_017008412.1 Intron XP_016863901.1

View Full Product Details