Product Details

SNP ID
rs12037405
Assay Type
Functionally tested
NCBI dbSNP Submissions
24
Location
Chr.1:70415464 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTAGGAAATGAAATACAGGTTAG[A/G]ATTAGGTTTATTCTGTTGGCTCATG
Phenotype
MIM: 607657
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTH PubMed Links

Gene Details

Gene
CTH
Gene Name
cystathionine gamma-lyase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190463.1 Intron NP_001177392.1
NM_001902.5 Intron NP_001893.2
NM_153742.4 Intron NP_714964.2
XM_005270509.3 Intron XP_005270566.1
XM_017000416.1 Intron XP_016855905.1

View Full Product Details