Product Details

SNP ID
rs11806447
Assay Type
Functionally tested
NCBI dbSNP Submissions
34
Location
Chr.1:71412215 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTCTTCACTCTAGTCATACAAGCC[A/G]TTTTCTGAGCACATAGAGTGTTTCA
Phenotype
MIM: 613173
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NEGR1 PubMed Links

Gene Details

Gene
NEGR1
Gene Name
neuronal growth regulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173808.2 Intron NP_776169.2
XM_011541200.2 Intron XP_011539502.1
XM_011541201.2 Intron XP_011539503.1
XM_017000961.1 Intron XP_016856450.1

View Full Product Details