Product Details

SNP ID
rs11205977
Assay Type
DME
NCBI dbSNP Submissions
24
Location
Chr.1:52602421 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
CTCCGGAACAAGCCATGGTGGCGGC[G/T]ACGGTGGCAGCGGCGTGGCTGCTCC
Phenotype
MIM: 615784
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GPX7 PubMed Links

Gene Details

Gene
GPX7
Gene Name
glutathione peroxidase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015696.4 51 Silent Mutation GCG,GCT A4A NP_056511.2

View Full Product Details