Product Details
- SNP ID
-
rs12143624
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
29
- Location
-
Chr.1:39000301 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACTTATCTCTGTACTGAATTCAAAT[C/T]TTCTGTGAGTGAACCTGATTAGGAG
- Phenotype
-
MIM: 615164
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
AKIRIN1
PubMed Links
Gene Details
- Gene
- AKIRIN1
- Gene Name
- akirin 1
View Full Product Details