Product Details

SNP ID
rs12748869
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:36799531 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGACATGGACCCCTTGGTGACA[A/T]GGCTTCTCCTTTTACGCATATTTGG
Phenotype
MIM: 138243
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GRIK3 PubMed Links

Gene Details

Gene
GRIK3
Gene Name
glutamate ionotropic receptor kainate type subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000831.3 5215 UTR 3 NP_000822.2

View Full Product Details