Product Details

SNP ID
rs12077489
Assay Type
Functionally tested
NCBI dbSNP Submissions
29
Location
Chr.1:28608390 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACTCAGGTGGTGGAGGTTGCAGAC[A/G]AAGGTTATTCTCTACGAATTGAACT
Phenotype
MIM: 600773
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TAF12 PubMed Links

Gene Details

Gene
TAF12
Gene Name
TATA-box binding protein associated factor 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135218.1 Intron NP_001128690.1
NM_005644.3 Intron NP_005635.1
XM_017002186.1 Intron XP_016857675.1

View Full Product Details