Product Details
- SNP ID
-
rs10934681
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:124731474 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAACAATGTATAGCTAATCAATCGC[C/T]TGTGTTATTATTTTATTTATTTATT
- Phenotype
-
MIM: 604605
MIM: 613891
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KALRN
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs17843787] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KALRN
- Gene Name
- kalirin, RhoGEF kinase
There are no transcripts associated with this gene.
- Gene
- MIR544B
- Gene Name
- microRNA 544b
There are no transcripts associated with this gene.
- Gene
- UMPS
- Gene Name
- uridine monophosphate synthetase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000373.3 |
|
Intron |
|
|
NP_000364.1 |
View Full Product Details