Product Details
- SNP ID
-
rs11066471
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:109390234 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTCTTTTTTTTTTTTTTGAGTTGGG[C/T]CTTGCTAAGTTGCCCAGGCTAGTCT
- Phenotype
-
MIM: 614636
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MYO1H
PubMed Links
Gene Details
- Gene
- MYO1H
- Gene Name
- myosin IH
View Full Product Details