Product Details

SNP ID
rs11715803
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:37989529 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGAACTGACTGCCGCTTGGTTGA[C/G]AGCGGGCATTGGTGACTTAGGAAAG
Phenotype
MIM: 608592
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CTDSPL PubMed Links

Gene Details

Gene
CTDSPL
Gene Name
CTD small phosphatase like
There are no transcripts associated with this gene.

Gene
VILL
Gene Name
villin like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015873.3 Intron NP_056957.3
XM_005265191.3 Intron XP_005265248.1
XM_005265192.4 Intron XP_005265249.1
XM_006713184.3 Intron XP_006713247.1
XM_011533770.2 Intron XP_011532072.1
XM_011533771.1 Intron XP_011532073.1
XM_011533773.2 Intron XP_011532075.1

View Full Product Details