Product Details

SNP ID
rs12079605
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:67417856 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATGAGGAGCAGAAAACAGAAACT[C/T]TGAGCTCTGTCAATAACACGGTAAC
Phenotype
MIM: 607378
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SERBP1 PubMed Links

Gene Details

Gene
SERBP1
Gene Name
SERPINE1 mRNA binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018067.1 Intron NP_001018077.1
NM_001018068.1 Intron NP_001018078.1
NM_001018069.1 Intron NP_001018079.1
NM_015640.3 Intron NP_056455.3

View Full Product Details