Product Details

SNP ID
rs10888976
Assay Type
Functionally tested
NCBI dbSNP Submissions
67
Location
Chr.1:56495682 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGCAGACCATGCCTTGCTCATTCC[C/T]GGGCCCTCACAGGAAATCCTGGTGG
Phenotype
MIM: 607125
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLPP3 PubMed Links

Gene Details

Gene
PLPP3
Gene Name
phospholipid phosphatase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003713.4 2373 UTR 3 NP_003704.3

View Full Product Details