Product Details

SNP ID
rs3918052
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:99489381 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTAATGAAGTTTTAAAATTTCGT[C/T]GGGGCCAGCCACTGTGGCTCATACC
Phenotype
MIM: 603544 MIM: 615671
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCNK PubMed Links

Gene Details

Gene
CCNK
Gene Name
cyclin K
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099402.1 Intron NP_001092872.1
XM_005268154.4 Intron XP_005268211.1
XM_011537275.2 Intron XP_011535577.1
Gene
SETD3
Gene Name
SET domain containing 3
There are no transcripts associated with this gene.

View Full Product Details