Product Details

SNP ID
rs12281343
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:94980271 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTATTATCTTCTAAAGGTTTCATA[A/G]CTGCACATGAAATTCATTTTTGTGC
Phenotype
MIM: 609766
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CWC15 PubMed Links

Gene Details

Gene
CWC15
Gene Name
CWC15 spliceosome-associated protein
There are no transcripts associated with this gene.

Gene
KDM4D
Gene Name
lysine demethylase 4D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018039.2 Intron NP_060509.2

View Full Product Details