Product Details
- SNP ID
-
rs11020995
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:94983834 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATCAAATGCACCTGGAGGTGTGGAG[A/C]AACAGAAACTCCTATTAGTACATAG
- Phenotype
-
MIM: 609766
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KDM4D
PubMed Links
Gene Details
- Gene
- KDM4D
- Gene Name
- lysine demethylase 4D
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018039.2 |
|
Intron |
|
|
NP_060509.2 |
View Full Product Details