Product Details

SNP ID
rs12285250
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:125452921 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGAGTCTCAAGGAAGCCTGAGGTG[G/T]TGAGTGCCAGAGCTCTAGATCAAAG
Phenotype
MIM: 604825
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FEZ1 PubMed Links

Gene Details

Gene
FEZ1
Gene Name
fasciculation and elongation protein zeta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005103.4 Intron NP_005094.1
NM_022549.3 Intron NP_072043.1
XM_005271734.2 Intron XP_005271791.1
XM_005271735.2 Intron XP_005271792.1

View Full Product Details