Product Details

SNP ID
rs10750303
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:125835304 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGGCACAGTGAGCTTGAGGAACC[C/T]AAAGGTGCGAGGGAGCCGGAGGGTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PATE4 PubMed Links

Gene Details

Gene
PATE4
Gene Name
prostate and testis expressed 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144874.1 Intron NP_001138346.1

View Full Product Details