Product Details
- SNP ID
-
rs4937054
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:125836916 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTTCAATTCAACATTGCTGAGTGAA[C/G]GAAAAAATGAATAAACTTTAGGGTT
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PATE4
PubMed Links
Gene Details
- Gene
- PATE4
- Gene Name
- prostate and testis expressed 4
View Full Product Details