Product Details
- SNP ID
-
rs12342266
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:21409867 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCATGCTATGAATTGAATCAAATG[G/T]GTCAAGTGTTTTCAGGAGTGTTAAG
- Phenotype
-
MIM: 147568
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
IFNA8
PubMed Links
Gene Details
- Gene
- IFNA8
- Gene Name
- interferon alpha 8
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002170.3 |
721 |
UTR 3 |
|
|
NP_002161.2 |
View Full Product Details