Product Details

SNP ID
rs13299803
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:22447760 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTTGGTCTTTGATTTACTCTTTG[C/T]TCAAGGAATGGTTGTTCTGGAAAGA
Phenotype
MIM: 614803
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DMRTA1 PubMed Links

Gene Details

Gene
DMRTA1
Gene Name
DMRT like family A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022160.2 Intron NP_071443.2

View Full Product Details