Product Details

SNP ID
rs10811179
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:19413753 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAGGACAGAAGAAAGTCAACTGAT[C/G]GCTGAAAAGAATCTTGAAGCAAAAT
Phenotype
MIM: 613492
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ACER2 PubMed Links

Gene Details

Gene
ACER2
Gene Name
alkaline ceramidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010887.2 Intron NP_001010887.2
XM_005251448.3 Intron XP_005251505.1
XM_011517858.1 Intron XP_011516160.1
XM_011517859.2 Intron XP_011516161.1
XM_017014694.1 Intron XP_016870183.1

View Full Product Details