Product Details

SNP ID
rs11233520
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:83195529 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATATAGATATCTCCCTTCTGGAAA[A/T]TTTCCTTGGGTACCTCAAGAGCAGC
Phenotype
MIM: 608876
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD42 PubMed Links

Gene Details

Gene
ANKRD42
Gene Name
ankyrin repeat domain 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300972.1 Intron NP_001287901.1
NM_001300973.1 Intron NP_001287902.1
NM_001300975.1 Intron NP_001287904.1
NM_001300976.1 Intron NP_001287905.1
NM_001300977.1 Intron NP_001287906.1
NM_182603.3 Intron NP_872409.2
Gene
LOC100506282
Gene Name
uncharacterized LOC100506282
There are no transcripts associated with this gene.

Gene
PCF11
Gene Name
PCF11 cleavage and polyadenylation factor subunit
There are no transcripts associated with this gene.

Gene
PCF11-AS1
Gene Name
PCF11 anstisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details