Product Details

SNP ID
rs13206144
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30944605 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCTCTTTCAAATGAGTGAACAAA[A/G]CAGATGGCGGGGACCTTTGGCACTG
Phenotype
MIM: 613928
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DPCR1 PubMed Links

Gene Details

Gene
DPCR1
Gene Name
diffuse panbronchiolitis critical region 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080870.3 Intron NP_543146.2

View Full Product Details