Product Details

SNP ID
rs13041150
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:3082642 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGAGCGCGGGGCTCAGTAGGCGTC[A/G]GGCTGGGCGGGCTCGAAGGGCTCGG
Phenotype
MIM: 192340
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AVP PubMed Links

Gene Details

Gene
AVP
Gene Name
arginine vasopressin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000490.4 533 Silent Mutation CCC,CCT P161P NP_000481.2
XM_011529267.1 533 Silent Mutation CCC,CCT P161P XP_011527569.1

View Full Product Details