Product Details

SNP ID
rs12351247
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:127430288 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAATTTCACCTTACTATTTTAGATT[C/T]GGGGGTATGTGTGCAGGGTTGTTAA
Phenotype
MIM: 194552
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF79 PubMed Links

Gene Details

Gene
ZNF79
Gene Name
zinc finger protein 79
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286696.1 Intron NP_001273625.1
NM_001286697.1 Intron NP_001273626.1
NM_001286698.1 Intron NP_001273627.1
NM_001322260.1 Intron NP_001309189.1
NM_007135.2 Intron NP_009066.2
XM_006717279.3 Intron XP_006717342.1

View Full Product Details