Product Details
- SNP ID
-
rs10840986
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:18244642 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCAGGAAAATACTACAATTTCCAG[A/C]CATTATTTAATCAGGTATGTCTTAG
- Phenotype
-
MIM: 609001
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PIK3C2G
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs34749004] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PIK3C2G
- Gene Name
- phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 gamma
View Full Product Details