Product Details

SNP ID
rs11050202
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:29336732 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTGTCCCTACTTTATTTGAAATA[C/T]TGATTCCAACCTGGAACTGCAAAAG
Phenotype
MIM: 612236 MIM: 616156
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ERGIC2 PubMed Links

Gene Details

Gene
ERGIC2
Gene Name
ERGIC and golgi 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016570.2 Intron NP_057654.2
Gene
FAR2
Gene Name
fatty acyl-CoA reductase 2
There are no transcripts associated with this gene.

View Full Product Details