Product Details

SNP ID
rs11872436
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:62191304 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCTGTCATTTTAGTGAGAATGTA[C/G]CAGTGTCTCATTGTGGCTTTAATTT
Phenotype
MIM: 606097
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KIAA1468 PubMed Links

Gene Details

Gene
KIAA1468
Gene Name
KIAA1468
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020854.3 Intron NP_065905.2
XM_005266730.2 Intron XP_005266787.1
XM_011526110.1 Intron XP_011524412.1
XM_011526111.1 Intron XP_011524413.1
XM_011526112.1 Intron XP_011524414.1
XM_011526114.1 Intron XP_011524416.1
XM_011526117.2 Intron XP_011524419.1
XM_017025882.1 Intron XP_016881371.1
XM_017025883.1 Intron XP_016881372.1
Gene
PIGN
Gene Name
phosphatidylinositol glycan anchor biosynthesis class N
There are no transcripts associated with this gene.

View Full Product Details