Product Details
- SNP ID
-
hCV31396346
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:91500883 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGCGCATCCGATCGACTCGGTAGG[C/T]GGGGATCTCTTGGAGACGGCGACCC
- Phenotype
-
MIM: 603311
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CDC7
PubMed Links
Gene Details
- Gene
- CDC7
- Gene Name
- cell division cycle 7
There are no transcripts associated with this gene.
View Full Product Details