Product Details

SNP ID
rs12835978
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:73452228 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTGGCTGGTGTTCCTGAATGTAA[A/G]GAGGGTTCAGAATGGGATCTGATGC
Phenotype
MIM: 300025
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDX4 PubMed Links

Gene Details

Gene
CDX4
Gene Name
caudal type homeobox 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005193.1 Intron NP_005184.1

View Full Product Details