Product Details
- SNP ID
-
rs13346156
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:12627279 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCTCTTGCAGAACATTGAGTATATT[C/T]GTTTTTAAACAAGTCAGACGGGGCA
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF791
PubMed Links
Gene Details
- Gene
- ZNF791
- Gene Name
- zinc finger protein 791
View Full Product Details