Product Details

SNP ID
rs12603900
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:62572577 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATACTAATTCATTTTGTATACTAT[G/T]CCCTAAGGAGTTCTTACTAGTTGGA
Phenotype
MIM: 608439
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TLK2 PubMed Links

Gene Details

Gene
TLK2
Gene Name
tousled like kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284333.1 Intron NP_001271262.1
NM_001284363.1 Intron NP_001271292.1
NM_006852.3 Intron NP_006843.2
XM_011524215.2 Intron XP_011522517.1
XM_011524216.2 Intron XP_011522518.1
XM_011524217.2 Intron XP_011522519.1
XM_011524218.2 Intron XP_011522520.1
XM_011524220.2 Intron XP_011522522.1
XM_011524221.2 Intron XP_011522523.1
XM_011524222.2 Intron XP_011522524.1
XM_011524223.2 Intron XP_011522525.1
XM_011524224.2 Intron XP_011522526.1
XM_011524225.2 Intron XP_011522527.1
XM_011524226.2 Intron XP_011522528.1
XM_011524227.2 Intron XP_011522529.1
XM_011524228.2 Intron XP_011522530.1
XM_011524229.2 Intron XP_011522531.1
XM_011524231.1 Intron XP_011522533.1
XM_017024044.1 Intron XP_016879533.1
XM_017024045.1 Intron XP_016879534.1
XM_017024046.1 Intron XP_016879535.1
XM_017024047.1 Intron XP_016879536.1
XM_017024048.1 Intron XP_016879537.1
XM_017024049.1 Intron XP_016879538.1
XM_017024050.1 Intron XP_016879539.1
XM_017024051.1 Intron XP_016879540.1
XM_017024052.1 Intron XP_016879541.1
XM_017024053.1 Intron XP_016879542.1
XM_017024054.1 Intron XP_016879543.1
XM_017024055.1 Intron XP_016879544.1
XM_017024056.1 Intron XP_016879545.1
XM_017024057.1 Intron XP_016879546.1
XM_017024058.1 Intron XP_016879547.1
XM_017024059.1 Intron XP_016879548.1
XM_017024060.1 Intron XP_016879549.1

View Full Product Details