Product Details

SNP ID
rs7236252
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:732476 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAAAACAAAACACCAATCACACT[A/G]CTGGGAAAGAACAGTTATTTAGGGT
Phenotype
MIM: 164880
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
YES1 PubMed Links

Gene Details

Gene
YES1
Gene Name
YES proto-oncogene 1, Src family tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005433.3 Intron NP_005424.1
XM_005258139.3 Intron XP_005258196.1
XM_017025960.1 Intron XP_016881449.1

View Full Product Details