Product Details

SNP ID
rs12319965
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:912867 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATAAATTATTTATATTAAATGAA[A/G]ATTGTAGCCTGGATTGATACAAAGT
Phenotype
MIM: 600392 MIM: 605232
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAD52 PubMed Links

Gene Details

Gene
RAD52
Gene Name
RAD52 homolog, DNA repair protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297419.1 2177 UTR 3 NP_001284348.1
NM_001297420.1 2177 Intron NP_001284349.1
NM_001297421.1 2177 UTR 3 NP_001284350.1
NM_001297422.1 2177 Intron NP_001284351.1
NM_134424.3 2177 UTR 3 NP_602296.2
XM_005253720.4 2177 UTR 3 XP_005253777.1
XM_005253721.2 2177 UTR 3 XP_005253778.1
XM_011520990.2 2177 UTR 3 XP_011519292.1
XM_011520991.2 2177 UTR 3 XP_011519293.1
XM_011520993.2 2177 Intron XP_011519295.1
XM_011520995.2 2177 UTR 3 XP_011519297.1
XM_017019769.1 2177 UTR 3 XP_016875258.1
XM_017019770.1 2177 UTR 3 XP_016875259.1
XM_017019771.1 2177 Intron XP_016875260.1
Gene
WNK1
Gene Name
WNK lysine deficient protein kinase 1
There are no transcripts associated with this gene.

View Full Product Details