Product Details
- SNP ID
-
rs12383583
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:104250272 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAATAACTGTTGATGACAGGGGCCC[A/G]TGTGATGTGTACAGGAGCCAGGCCA
- Phenotype
-
MIM: 300154
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ESX1
PubMed Links
Gene Details
- Gene
- ESX1
- Gene Name
- ESX homeobox 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_153448.3 |
1246 |
Missense Mutation |
AGG,GGG |
R393G |
NP_703149.1 |
View Full Product Details