Product Details

SNP ID
rs12024799
Assay Type
Functionally tested
NCBI dbSNP Submissions
29
Location
Chr.1:161103213 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGAATTGGAATTTTGCAGACAAAT[G/T]GGTATAGAAGACAGCTGGAGTTAAG
Phenotype
MIM: 613466
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KLHDC9 PubMed Links

Gene Details

Gene
KLHDC9
Gene Name
kelch domain containing 9
There are no transcripts associated with this gene.

Gene
PFDN2
Gene Name
prefoldin subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012394.3 Intron NP_036526.2
XM_011509624.2 Intron XP_011507926.1

View Full Product Details