Product Details

SNP ID
rs6656542
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:160945931 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTGGAGAATATTTTCACGACCTC[A/G]GTGTAAGGAGGTACATCTTAAAGAA
Phenotype
MIM: 609874
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITLN2 PubMed Links

Gene Details

Gene
ITLN2
Gene Name
intelectin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080878.2 Intron NP_543154.1
XM_011509210.1 Intron XP_011507512.1
XM_011509211.1 Intron XP_011507513.1
Gene
LOC101928372
Gene Name
uncharacterized LOC101928372
There are no transcripts associated with this gene.

View Full Product Details