Product Details

SNP ID
rs11584916
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:156700970 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACGGTGGTGGAGGTTTTGATGTA[A/G]AAAGTGTCTCCCTCCTGTTTGATCT
Phenotype
MIM: 180231
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRABP2 PubMed Links

Gene Details

Gene
CRABP2
Gene Name
cellular retinoic acid binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199723.1 374 Silent Mutation TTC,TTT F51F NP_001186652.1
NM_001878.3 374 Silent Mutation TTC,TTT F51F NP_001869.1
XM_017000343.1 374 Silent Mutation TTC,TTT F51F XP_016855832.1

View Full Product Details