Product Details

SNP ID
rs12041712
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:156616612 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATTTATATCTACTTCTCCCTCTGC[C/T]GGAGGCTGTTTGTTTGAGTTGGCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HAPLN2 PubMed Links

Gene Details

Gene
HAPLN2
Gene Name
hyaluronan and proteoglycan link protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021817.2 Intron NP_068589.1
XM_005245415.2 Intron XP_005245472.1
XM_011509853.2 Intron XP_011508155.1
XM_017002020.1 Intron XP_016857509.1
XM_017002021.1 Intron XP_016857510.1
Gene
LOC101928177
Gene Name
uncharacterized LOC101928177
There are no transcripts associated with this gene.

View Full Product Details