Product Details
- SNP ID
-
rs7169606
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:42279305 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACCCACGTGCTTTCTAACACTGTTA[A/C]ACAATGCCCATTTGCATGTTTAGCC
- Phenotype
-
MIM: 104180
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GANC
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77082056] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GANC
- Gene Name
- glucosidase alpha, neutral C
- Gene
- TMEM87A
- Gene Name
- transmembrane protein 87A
There are no transcripts associated with this gene.
View Full Product Details