Product Details

SNP ID
rs4305190
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44389596 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCAATACACTTCGATAAATTCTT[C/T]TGTTTTCTGTTCTATTAATCTATTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF285 PubMed Links

Gene Details

Gene
ZNF285
Gene Name
zinc finger protein 285
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291488.1 Intron NP_001278417.1
NM_001291489.1 Intron NP_001278418.1
NM_001291490.1 Intron NP_001278419.1
NM_001291491.1 Intron NP_001278420.1
NM_152354.5 Intron NP_689567.4

View Full Product Details