Product Details

SNP ID
rs12069017
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:10947730 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCTCAGGAGGCCTGTGGCTTTCA[A/G]CACTGAGGGCCAAGGTGGGCTCTGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf127 PubMed Links

Gene Details

Gene
C1orf127
Gene Name
chromosome 1 open reading frame 127
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170754.1 2323 Missense Mutation GCT,GTT A802V NP_001164225.1
XM_017000373.1 2323 Missense Mutation GCT,GTT A766V XP_016855862.1

View Full Product Details