Product Details
- SNP ID
-
rs12556694
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:43658819 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATGAAAGTAGTTGCAAGTGTGGCT[C/T]GTGTTAGATCCTGGACATAAACTGA
- Phenotype
-
MIM: 309850
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MAOA
PubMed Links
Gene Details
- Gene
- MAOA
- Gene Name
- monoamine oxidase A
View Full Product Details