Product Details

SNP ID
rs10305287
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40447415 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGCGTTTGGACGGGGAGTTCCTA[A/C]GCTGGGCTCTCAGTCCCTTTGGGAC
Phenotype
MIM: 146733
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IGFBP4 PubMed Links

Gene Details

Gene
IGFBP4
Gene Name
insulin like growth factor binding protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001552.2 Intron NP_001543.2

View Full Product Details