Product Details

SNP ID
rs12061338
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:15861483 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGTCACTAGTTAAGTGAAATATTA[A/G]AAATATTTGTATATTTCCCATAGTT
Phenotype
MIM: 613484
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPEN PubMed Links

Gene Details

Gene
SPEN
Gene Name
spen family transcriptional repressor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015001.2 Intron NP_055816.2

View Full Product Details