Product Details

SNP ID
rs10993328
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:94726763 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCGGCTGAGACAGGTAACCTGTTC[G/T]CTCCCTGTTGGGTCCCTGCCTTCCT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C9orf3 PubMed Links

Gene Details

Gene
C9orf3
Gene Name
chromosome 9 open reading frame 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193329.1 Intron NP_001180258.1
NM_001193331.2 Intron NP_001180260.1
NM_032823.5 Intron NP_116212.3
XM_005252284.2 Intron XP_005252341.1
XM_006717306.1 Intron XP_006717369.1
XM_011519119.1 Intron XP_011517421.1
XM_011519120.2 Intron XP_011517422.1
XM_011519121.2 Intron XP_011517423.1
XM_011519122.2 Intron XP_011517424.1
XM_011519123.2 Intron XP_011517425.1
XM_011519127.1 Intron XP_011517429.1
XM_011519129.1 Intron XP_011517431.1
XM_011519130.1 Intron XP_011517432.1
XM_011519132.1 Intron XP_011517434.1
XM_011519134.2 Intron XP_011517436.1
XM_017015229.1 Intron XP_016870718.1
XM_017015230.1 Intron XP_016870719.1
XM_017015231.1 Intron XP_016870720.1
XM_017015232.1 Intron XP_016870721.1
XM_017015233.1 Intron XP_016870722.1
XM_017015234.1 Intron XP_016870723.1
XM_017015235.1 Intron XP_016870724.1
XM_017015236.1 Intron XP_016870725.1
XM_017015237.1 Intron XP_016870726.1

View Full Product Details