Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001193329.1 | Intron | NP_001180258.1 | |||
NM_001193331.2 | Intron | NP_001180260.1 | |||
NM_032823.5 | Intron | NP_116212.3 | |||
XM_005252284.2 | Intron | XP_005252341.1 | |||
XM_006717306.1 | Intron | XP_006717369.1 | |||
XM_011519119.1 | Intron | XP_011517421.1 | |||
XM_011519120.2 | Intron | XP_011517422.1 | |||
XM_011519121.2 | Intron | XP_011517423.1 | |||
XM_011519122.2 | Intron | XP_011517424.1 | |||
XM_011519123.2 | Intron | XP_011517425.1 | |||
XM_011519127.1 | Intron | XP_011517429.1 | |||
XM_011519129.1 | Intron | XP_011517431.1 | |||
XM_011519130.1 | Intron | XP_011517432.1 | |||
XM_011519132.1 | Intron | XP_011517434.1 | |||
XM_011519134.2 | Intron | XP_011517436.1 | |||
XM_017015229.1 | Intron | XP_016870718.1 | |||
XM_017015230.1 | Intron | XP_016870719.1 | |||
XM_017015231.1 | Intron | XP_016870720.1 | |||
XM_017015232.1 | Intron | XP_016870721.1 | |||
XM_017015233.1 | Intron | XP_016870722.1 | |||
XM_017015234.1 | Intron | XP_016870723.1 | |||
XM_017015235.1 | Intron | XP_016870724.1 | |||
XM_017015236.1 | Intron | XP_016870725.1 | |||
XM_017015237.1 | Intron | XP_016870726.1 |