Product Details

SNP ID
rs10788814
Assay Type
Functionally tested
NCBI dbSNP Submissions
45
Location
Chr.1:151881278 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACTCTTGGGCTCTAGCGATCTTC[C/T]TGCATCAACCTCCCAAGTAGCTAGG
Phenotype
MIM: 606388
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
THEM4 PubMed Links

Gene Details

Gene
THEM4
Gene Name
thioesterase superfamily member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_053055.4 Intron NP_444283.2
XM_011509142.2 Intron XP_011507444.1

View Full Product Details