Product Details

SNP ID
rs11205363
Assay Type
Functionally tested
NCBI dbSNP Submissions
28
Location
Chr.1:150371133 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGTTTTGCAGAGAATTTACAAAGT[A/G]CGGTTAATGTATGCCAATGGTTTCT
Phenotype
MIM: 614695
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPRD2 PubMed Links

Gene Details

Gene
RPRD2
Gene Name
regulation of nuclear pre-mRNA domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297673.1 Intron NP_001284602.1
NM_001297674.1 Intron NP_001284603.1
NM_015203.4 Intron NP_056018.2
XM_005245033.2 Intron XP_005245090.1
XM_006711240.2 Intron XP_006711303.1
XM_011509344.2 Intron XP_011507646.1
XM_017000763.1 Intron XP_016856252.1
XM_017000764.1 Intron XP_016856253.1
XM_017000765.1 Intron XP_016856254.1
XM_017000766.1 Intron XP_016856255.1
XM_017000767.1 Intron XP_016856256.1

View Full Product Details