Product Details

SNP ID
rs12974634
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41836190 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTATGGCCAGAGAAAAGAATCAC[A/C]CTGTTCTCCTTTCCTAGGAACCCAG
Phenotype
MIM: 614806
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DMRTC2 PubMed Links

Gene Details

Gene
DMRTC2
Gene Name
DMRT like family C2
There are no transcripts associated with this gene.

Gene
LYPD4
Gene Name
LY6/PLAUR domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291419.1 Intron NP_001278348.1
NM_173506.6 Intron NP_775777.3
XM_005258543.3 Intron XP_005258600.1
XM_005278383.4 Intron XP_005278440.1
XM_005278385.4 Intron XP_005278442.1
XM_005278387.4 Intron XP_005278444.1
XM_005278388.3 Intron XP_005278445.1
XM_006723035.2 Intron XP_006723098.1
XM_011526483.2 Intron XP_011524785.1
XM_011526484.2 Intron XP_011524786.1

View Full Product Details